Product Details

SNP ID
rs1131431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:139372697 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTGTGCAGTAATATCTGCCCTTC[A/G]AAGTTCATGCAACCAACTAATGCAA
Phenotype
MIM: 602937
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CITED2 PubMed Links

Gene Details

Gene
CITED2
Gene Name
Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168388.2 1494 UTR 3 NP_001161860.1
NM_001168389.2 1494 UTR 3 NP_001161861.2
NM_006079.4 1494 UTR 3 NP_006070.2

View Full Product Details