Product Details

SNP ID
rs1475931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113109612 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCACAGCGTCCACCTGAGGCCTC[G/T]TCCTCCCAGCAAAGGTCGTCCCTCC
Phenotype
MIM: 613878 MIM: 609499
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
F7 PubMed Links

Gene Details

Gene
F7
Gene Name
coagulation factor VII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000131.4 Intron NP_000122.1
NM_001267554.1 Intron NP_001254483.1
NM_019616.3 Intron NP_062562.1
XM_006719963.3 Intron XP_006720026.2
XM_011537474.2 Intron XP_011535776.2
XM_011537475.2 Intron XP_011535777.2
XM_011537476.2 Intron XP_011535778.1
Gene
MCF2L
Gene Name
MCF.2 cell line derived transforming sequence like
There are no transcripts associated with this gene.

View Full Product Details