Product Details
- SNP ID
-
rs1168308
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:66348403 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATTTGTTGCATAGGGAATAGCATAC[A/G]TCTTAGTTAAAATCATTCCTATACG
- Phenotype
-
MIM: 604597
MIM: 614539
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GRIP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138816960,rs200677568] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GRIP1
- Gene Name
- glutamate receptor interacting protein 1
- Gene
- HELB
- Gene Name
- DNA helicase B
There are no transcripts associated with this gene.
View Full Product Details