Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001287214.1 | 38 | Intron | NP_001274143.1 | ||
NM_001287215.1 | 38 | Intron | NP_001274144.1 | ||
NM_001287216.1 | 38 | Intron | NP_001274145.1 | ||
NM_001634.5 | 38 | Intron | NP_001625.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001300960.1 | 38 | Intron | NP_001287889.1 | ||
NM_001300963.1 | 38 | UTR 5 | NP_001287892.1 | ||
NM_001300964.1 | 38 | Intron | NP_001287893.1 | ||
NM_015076.4 | 38 | Intron | NP_055891.1 | ||
XM_005266871.3 | 38 | Intron | XP_005266928.1 | ||
XM_011535630.2 | 38 | Intron | XP_011533932.1 | ||
XM_011535631.2 | 38 | Intron | XP_011533933.1 | ||
XM_011535632.1 | 38 | Intron | XP_011533934.1 | ||
XM_017010587.1 | 38 | Intron | XP_016866076.1 | ||
XM_017010588.1 | 38 | Intron | XP_016866077.1 |