Product Details

SNP ID
rs1054797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41354854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGGCCCACCACCCACGAAAGCC[C/T]GTGCCAACTGTTCTCGCCAACTGCC
Phenotype
MIM: 611951 MIM: 190180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B9D2 PubMed Links

Gene Details

Gene
B9D2
Gene Name
B9 protein domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030578.3 594 Missense Mutation CAG,CGG Q125R NP_085055.2
XM_011527349.2 594 Missense Mutation CAG,CGG Q125R XP_011525651.1
XM_011527350.1 594 Missense Mutation CAG,CGG Q72R XP_011525652.1
Gene
TGFB1
Gene Name
transforming growth factor beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011527242.1 594 Intron XP_011525544.1
Gene
TMEM91
Gene Name
transmembrane protein 91
There are no transcripts associated with this gene.

View Full Product Details