Product Details

SNP ID
rs1495276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:60397787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCGGGGAGGGGGTGTTTCTGC[A/G]GCGGGATCGGGAGGCGAGGGCTGGG
Phenotype
MIM: 614392
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TDRD3 PubMed Links

Gene Details

Gene
TDRD3
Gene Name
tudor domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146070.1 Intron NP_001139542.1
NM_001146071.1 Intron NP_001139543.1
NM_030794.2 Intron NP_110421.1
XM_005266556.4 Intron XP_005266613.1
XM_005266560.2 Intron XP_005266617.1
XM_011535247.1 Intron XP_011533549.1
XM_011535249.2 Intron XP_011533551.1
XM_017020777.1 Intron XP_016876266.1

View Full Product Details