Product Details

SNP ID
rs1054383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38305168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACAGCGGCCACGCCCTGGGGCGG[G/T]GGCCTGTGCAGCTGGAGATCTCTCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C19orf33 PubMed Links

Gene Details

Gene
C19orf33
Gene Name
chromosome 19 open reading frame 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317801.1 1160 Intron NP_001304730.1
NM_033520.2 1160 Intron NP_277055.1
Gene
YIF1B
Gene Name
Yip1 interacting factor homolog B, membrane trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039671.2 1160 UTR 3 NP_001034760.1
NM_001039672.2 1160 UTR 3 NP_001034761.1
NM_001039673.2 1160 UTR 3 NP_001034762.1
NM_001145461.1 1160 UTR 3 NP_001138933.1
NM_001145462.1 1160 UTR 3 NP_001138934.1
NM_001145463.1 1160 UTR 3 NP_001138935.1
XM_005259385.3 1160 UTR 3 XP_005259442.1
XM_017027449.1 1160 Intron XP_016882938.1
XM_017027450.1 1160 UTR 3 XP_016882939.1
XM_017027451.1 1160 UTR 3 XP_016882940.1

View Full Product Details