Product Details

SNP ID
rs1661179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44541314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCATTTTACAGATTGGGAAACT[C/G]AGGCCCAGGGAAAGGAAGCGTCTGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM22P PubMed Links

Gene Details

Gene
CEACAM22P
Gene Name
carcinoembryonic antigen related cell adhesion molecule 22, pseudogene
There are no transcripts associated with this gene.

View Full Product Details