Product Details

SNP ID
rs1056335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52415728 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATTTACATTGCACTGTGAAGTTC[T/C]AATAAATTCTCAATTGATCGGGGTT
Phenotype
MIM: 616821 MIM: 610903
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
THSD1 PubMed Links

Gene Details

Gene
THSD1
Gene Name
thrombospondin type 1 domain containing 1
There are no transcripts associated with this gene.

Gene
VPS36
Gene Name
vacuolar protein sorting 36 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282168.1 1481 UTR 3 NP_001269097.1
NM_001282169.1 1481 UTR 3 NP_001269098.1
NM_016075.3 1481 UTR 3 NP_057159.2

View Full Product Details