Product Details

SNP ID
rs1234256
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37226455 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATCTCCATTATGATGTCATCCAC[A/G]TTAATTCCATTGCTCTAAAAGGCCT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF383 PubMed Links

Gene Details

Gene
ZNF383
Gene Name
zinc finger protein 383
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152604.1 Intron NP_689817.1
XM_005258585.2 Intron XP_005258642.1
XM_005258587.2 Intron XP_005258644.1
XM_005258588.2 Intron XP_005258645.1
XM_011526586.2 Intron XP_011524888.1
XM_011526587.2 Intron XP_011524889.1
XM_011526588.2 Intron XP_011524890.1
XM_011526589.2 Intron XP_011524891.1
XM_011526590.2 Intron XP_011524892.1
XM_017026422.1 Intron XP_016881911.1
XM_017026423.1 Intron XP_016881912.1
XM_017026424.1 Intron XP_016881913.1

View Full Product Details