Product Details

SNP ID
rs1008910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22712955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTCGGTGTCAGTGGCCCCAGGA[A/C]AGACGGCCAGGATTACCTGTGGGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGLV3-19 PubMed Links

Gene Details

Gene
IGLV3-19
Gene Name
immunoglobulin lambda variable 3-19
There are no transcripts associated with this gene.

Gene
IGLV3-21
Gene Name
immunoglobulin lambda variable 3-21
There are no transcripts associated with this gene.

View Full Product Details