Product Details

SNP ID
rs1794735
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:120453513 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTAACTTATCTTACAGAACCCTGA[C/T]TGGGAAAGACAGGTGGTGAGGTAGA
Phenotype
MIM: 601943 MIM: 614943
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GATC PubMed Links

Gene Details

Gene
GATC
Gene Name
glutamyl-tRNA amidotransferase subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176818.2 Intron NP_789788.1
Gene
SRSF9
Gene Name
serine and arginine rich splicing factor 9
There are no transcripts associated with this gene.

Gene
TRIAP1
Gene Name
TP53 regulated inhibitor of apoptosis 1
There are no transcripts associated with this gene.

View Full Product Details