Product Details

SNP ID
rs917947
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:46353706 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACCCAGCCCAGGGCCTGTCCTCGCA[A/G]GGTTGGAGCACTGCCTCATGCCTCT
Phenotype
MIM: 600658
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP5C PubMed Links

Gene Details

Gene
PPP5C
Gene Name
protein phosphatase 5 catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204284.1 Intron NP_001191213.1
NM_006247.3 Intron NP_006238.1
XM_017026935.1 Intron XP_016882424.1

View Full Product Details