Product Details

SNP ID
rs698705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47434209 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGGTGGCTGCTATTACCGCATC[A/G]CTATGTCAGTTGAGGCTTCTGCCTG
Phenotype
MIM: 601901
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC8A2 PubMed Links

Gene Details

Gene
SLC8A2
Gene Name
solute carrier family 8 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015063.2 Intron NP_055878.1
XM_005259172.1 Intron XP_005259229.1
XM_017027159.1 Intron XP_016882648.1

View Full Product Details