Product Details
- SNP ID
-
rs1291350
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:12932606 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATAAAGCCCAGGTGGAAACTACAA[C/T]GAAATGTTCCATTTGTAGCATATAC
- Phenotype
-
MIM: 607437
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM234B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77563944] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM234B
- Gene Name
- family with sequence similarity 234 member B
- Gene
- GPRC5D
- Gene Name
- G protein-coupled receptor class C group 5 member D
There are no transcripts associated with this gene.
View Full Product Details