Product Details

SNP ID
rs36614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54201751 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTAACTTTTAGTGGCACTTGTG[A/G]AGTAGGAAAAGTGTATCTGGATCAG
Phenotype
MIM: 603631 MIM: 608754
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPS9 PubMed Links

Gene Details

Gene
RPS9
Gene Name
ribosomal protein S9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013.3 Intron NP_001004.2
NM_001321701.1 Intron NP_001308630.1
NM_001321702.1 Intron NP_001308631.1
NM_001321704.1 Intron NP_001308633.1
NM_001321705.1 Intron NP_001308634.1
NM_001321706.1 Intron NP_001308635.1
XM_017027112.1 Intron XP_016882601.1
Gene
TSEN34
Gene Name
tRNA splicing endonuclease subunit 34
There are no transcripts associated with this gene.

View Full Product Details