Product Details

SNP ID
rs1054807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:89519642 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTAATTAAAATTGAGACCCTAAA[C/T]CAACTAAGACTATACAACTTAAAAA
Phenotype
MIM: 603565 MIM: 614784
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALNT4 PubMed Links

Gene Details

Gene
GALNT4
Gene Name
polypeptide N-acetylgalactosaminyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003774.4 5165 UTR 3 NP_003765.2
Gene
POC1B
Gene Name
POC1 centriolar protein B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199777.1 5165 Intron NP_001186706.1
NM_172240.2 5165 Intron NP_758440.1
Gene
POC1B-GALNT4
Gene Name
POC1B-GALNT4 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199781.1 5165 UTR 3 NP_001186710.1
NM_001199782.1 5165 UTR 3 NP_001186711.1

View Full Product Details