Product Details
- SNP ID
-
rs1460026
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:28048441 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AAACATTGCCCTTTCAAGCAAGTTA[A/G]AACAATATCCCTCAACATTAAATAC
- Phenotype
-
MIM: 609476
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NLK
PubMed Links
Gene Details
- Gene
- NLK
- Gene Name
- nemo like kinase
View Full Product Details