Product Details
- SNP ID
-
rs1043721
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:29120465 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTTTCTGTGGCCTGGTGTGTTACAA[A/G]CATTTAATGTGGATCCTCATCTCCA
- Phenotype
-
MIM: 602712
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APBA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112042657,rs73372319] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APBA2
- Gene Name
- amyloid beta precursor protein binding family A member 2
There are no transcripts associated with this gene.
- Gene
- FAM189A1
- Gene Name
- family with sequence similarity 189 member A1
View Full Product Details