Product Details

SNP ID
rs4029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55044477 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCATGCCGTGCAGAGCCTGAACTC[A/G]GGATGACACCTGCACCTGCTCTCTG
Phenotype
MIM: 605546
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GP6 PubMed Links

Gene Details

Gene
GP6
Gene Name
glycoprotein VI platelet
There are no transcripts associated with this gene.

Gene
RDH13
Gene Name
retinol dehydrogenase 13 (all-trans/9-cis)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145971.1 1778 UTR 3 NP_001139443.1
NM_138412.3 1778 UTR 3 NP_612421.1
XM_005258473.1 1778 UTR 3 XP_005258530.1
XM_011526407.1 1778 UTR 3 XP_011524709.1
XM_011526408.2 1778 Intron XP_011524710.1

View Full Product Details