Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001145457.2 | Intron | NP_001138929.2 | |||
NM_001145458.2 | Intron | NP_001138930.2 | |||
NM_001242356.2 | Intron | NP_001229285.1 | |||
NM_001242357.2 | Intron | NP_001229286.1 | |||
NM_004829.6 | Intron | NP_004820.2 | |||
XM_011527528.2 | Intron | XP_011525830.1 | |||
XM_011527529.2 | Intron | XP_011525831.1 | |||
XM_011527530.2 | Intron | XP_011525832.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001127255.1 | Intron | NP_001120727.1 | |||
NM_139176.3 | Intron | NP_631915.2 | |||
NM_206828.3 | Intron | NP_996611.2 | |||
XM_006723075.3 | Intron | XP_006723138.1 | |||
XM_006723076.3 | Intron | XP_006723139.1 | |||
XM_011526596.2 | Intron | XP_011524898.1 | |||
XM_011526599.2 | Intron | XP_011524901.1 | |||
XM_011526601.2 | Intron | XP_011524903.1 |