Product Details

SNP ID
rs1509124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6778355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTTGCACAGACGAATAATGTCA[G/T]TCCTGAATTGGGAAATGTGATTCAC
Phenotype
MIM: 609106 MIM: 606717
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO39 PubMed Links

Gene Details

Gene
FBXO39
Gene Name
F-box protein 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153230.2 Intron NP_694962.1
XM_011523697.1 Intron XP_011521999.1
XM_017024283.1 Intron XP_016879772.1
Gene
XAF1
Gene Name
XIAP associated factor 1
There are no transcripts associated with this gene.

View Full Product Details