Product Details

SNP ID
rs1650983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53050822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCCCCCTTGGATGCCAGTGGGC[G/A]AAGATTCCGGGAAACTATGGAGGAA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ERVV-2 PubMed Links

Gene Details

Gene
ERVV-2
Gene Name
endogenous retrovirus group V member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191055.1 2177 Missense Mutation CAA,CGA Q524R NP_001177984.1

View Full Product Details