Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001320653.1 | 246 | Intron | NP_001307582.1 | ||
NM_002532.5 | 246 | Intron | NP_002523.2 | ||
XM_011523893.2 | 246 | Intron | XP_011522195.1 | ||
XM_017024694.1 | 246 | Intron | XP_016880183.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001033002.3 | 246 | UTR 5 | NP_001028174.2 | ||
NM_001160243.1 | 246 | UTR 5 | NP_001153715.1 | ||
NM_001160244.1 | 246 | UTR 5 | NP_001153716.1 | ||
NM_001160246.1 | 246 | UTR 5 | NP_001153718.1 | ||
NM_001160266.1 | 246 | UTR 5 | NP_001153738.1 |