Product Details

SNP ID
rs708726
Assay Type
Functionally tested
NCBI dbSNP Submissions
69
Location
Chr.1:205793011 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TCTCTCCAAGGAAGCCAGAGATGAC[G/T]CTTCACATTCTCCCTTGCTCTGCAC
Phenotype
MIM: 610801
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC41A1 PubMed Links

Gene Details

Gene
SLC41A1
Gene Name
solute carrier family 41 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173854.5 Intron NP_776253.3
XM_005245069.2 Intron XP_005245126.1

View Full Product Details