Product Details

SNP ID
rs1135812
Assay Type
Functionally tested
NCBI dbSNP Submissions
56
Location
Chr.1:45629600 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGCTCTGTCTTCATGTGGAACTC[C/T]TTGAGCTCATCCTCTGTGAGGGGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC17 PubMed Links

Gene Details

Gene
CCDC17
Gene Name
coiled-coil domain containing 17
There are no transcripts associated with this gene.

Gene
GPBP1L1
Gene Name
GC-rich promoter binding protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021639.4 2511 Silent Mutation NP_067652.1
XM_017002019.1 2511 Missense Mutation XP_016857508.1

View Full Product Details