Product Details

SNP ID
rs8450
Assay Type
Validated
NCBI dbSNP Submissions
46
Location
Chr.1:153947810 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TGCCCACCCCTAGGCATCCGGAAAA[A/G]CCCTGCTTCCAGGGCTCCCCTCTAC
Phenotype
MIM: 608972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRTC2 PubMed Links

Gene Details

Gene
CRTC2
Gene Name
CREB regulated transcription coactivator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181715.2 2540 UTR 3 NP_859066.1
XM_005244946.1 2540 UTR 3 XP_005245003.1
XM_005244949.2 2540 UTR 3 XP_005245006.1
XM_017000575.1 2540 UTR 3 XP_016856064.1
XM_017000576.1 2540 UTR 3 XP_016856065.1
XM_017000577.1 2540 UTR 3 XP_016856066.1
XM_017000578.1 2540 UTR 3 XP_016856067.1
XM_017000579.1 2540 UTR 3 XP_016856068.1
Gene
DENND4B
Gene Name
DENN domain containing 4B
There are no transcripts associated with this gene.

View Full Product Details