Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001105519.2 | Intron | NP_001098989.1 | |||
NM_001322426.1 | Intron | NP_001309355.1 | |||
XM_011532810.2 | Intron | XP_011531112.1 | |||
XM_011532811.2 | Intron | XP_011531113.1 | |||
XM_017003962.1 | Intron | XP_016859451.1 | |||
XM_017003963.1 | Intron | XP_016859452.1 | |||
XM_017003964.1 | Intron | XP_016859453.1 | |||
XM_017003965.1 | Intron | XP_016859454.1 | |||
XM_017003966.1 | Intron | XP_016859455.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001029881.2 | Intron | NP_001025052.1 | |||
XM_011532514.2 | Intron | XP_011530816.1 | |||
XM_011532517.2 | Intron | XP_011530819.1 | |||
XM_017003329.1 | Intron | XP_016858818.1 | |||
XM_017003330.1 | Intron | XP_016858819.1 | |||
XM_017003331.1 | Intron | XP_016858820.1 |