Product Details

SNP ID
rs1339848
Assay Type
Functionally Tested
NCBI dbSNP Submissions
48
Location
Chr.1:247948485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACAAATGTTGATCTCATCAAAT[A/G]TGAAACTCTGGGTAAGAAAATAAGA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR2L13 PubMed Links
Additional Information
For this assay, SNP(s) [rs140805913] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR2L13
Gene Name
olfactory receptor family 2 subfamily L member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304535.1 Intron NP_001291464.1
NM_175911.3 Intron NP_787107.1
XM_011544169.2 Intron XP_011542471.1
Gene
OR2L8
Gene Name
olfactory receptor family 2 subfamily L member 8 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001963.1 Intron NP_001001963.1

View Full Product Details