Product Details

SNP ID
rs1046863
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:154924787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCACACTGGGGAGCTCTGGGTT[C/T]TTGCCCAGAACAAGGGGCTGAGAAC
Phenotype
MIM: 607622
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PMVK PubMed Links

Gene Details

Gene
PMVK
Gene Name
phosphomevalonate kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323011.1 1244 UTR 3 NP_001309940.1
NM_001323012.1 1244 UTR 3 NP_001309941.1
NM_006556.3 1244 UTR 3 NP_006547.1

View Full Product Details