Product Details
- SNP ID
-
rs1035586
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:24177517 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTAAGCTTTTTAGTGCTTTAAAAC[A/G]TAATTTAATAATATGCTATCAGGGA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM228A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199984527] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM228A
- Gene Name
- family with sequence similarity 228 member A
- Gene
- FAM228B
- Gene Name
- family with sequence similarity 228 member B
There are no transcripts associated with this gene.
View Full Product Details