Product Details

SNP ID
rs840599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:187346569 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTTAATTTAAAAAATTAAGTGT[A/C]CCATCTTCACATTAGGAAATGTATT
Phenotype
MIM: 114190
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CALCRL PubMed Links

Gene Details

Gene
CALCRL
Gene Name
calcitonin receptor like receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271751.1 Intron NP_001258680.1
NM_005795.5 Intron NP_005786.1
XM_005246231.3 Intron XP_005246288.2
XM_005246232.3 Intron XP_005246289.2
XM_005246234.3 Intron XP_005246291.2
XM_017003151.1 Intron XP_016858640.1

View Full Product Details