Product Details

SNP ID
hCV8728190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24264684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAACTGCTCTCAAACCAACAAA[C/T]AGGCTTCTCTTGGCAACAGACTCAT
Phenotype
MIM: 614140
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POM121L9P PubMed Links

Gene Details

Gene
POM121L9P
Gene Name
POM121 transmembrane nucleoporin like 9, pseudogene
There are no transcripts associated with this gene.

Gene
SPECC1L
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1 like
There are no transcripts associated with this gene.

Gene
SPECC1L-ADORA2A
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details