Product Details

SNP ID
rs1044777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41848632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTACAATGTCATGTCTCAAGG[G/T]GTTCTAAATGCTACATCTCCTCTGA
Phenotype
MIM: 608778
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL10 PubMed Links

Gene Details

Gene
KLHL10
Gene Name
kelch like family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011524706.2 7296 Intron XP_011523008.1
Gene
KLHL11
Gene Name
kelch like family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018143.2 7296 UTR 3 NP_060613.1

View Full Product Details