Product Details

SNP ID
rs1061843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41802092 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCAGCCAGCAGCTGGAAACTCCT[C/T]GTCCAGGAGACTGTCCAGGTGAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P3H4 PubMed Links

Gene Details

Gene
P3H4
Gene Name
prolyl 3-hydroxylase family member 4 (non-enzymatic)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006455.2 2736 Intron NP_006446.1
XM_006721640.3 2736 UTR 3 XP_006721703.1

View Full Product Details