Product Details

SNP ID
rs13816
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:68021592 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGATTTGTCAAAATCAGGTTGCTTG[C/G]GGCAAAGGCTCTTTCACCGAGGATG
Phenotype
MIM: 607618 MIM: 609707
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDX28 PubMed Links

Gene Details

Gene
DDX28
Gene Name
DEAD-box helicase 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018380.3 2276 Silent Mutation CCC,CCG P537P NP_060850.2
Gene
DUS2
Gene Name
dihydrouridine synthase 2
There are no transcripts associated with this gene.

Gene
LOC100131303
Gene Name
uncharacterized LOC100131303
There are no transcripts associated with this gene.

View Full Product Details