Product Details

SNP ID
rs1212490
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67649433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGACTCCAGCCATCAATGGCTT[G/T]CTCTTAACCCCAGCCTCTTCAGTCT
Phenotype
MIM: 609377 MIM: 610859
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ACD PubMed Links

Gene Details

Gene
ACD
Gene Name
adrenocortical dysplasia homolog
There are no transcripts associated with this gene.

Gene
CARMIL2
Gene Name
capping protein regulator and myosin 1 linker 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013838.1 Intron NP_001013860.1
NM_001317026.1 Intron NP_001303955.1
XM_011522874.1 Intron XP_011521176.1
XM_011522875.2 Intron XP_011521177.2
XM_017022953.1 Intron XP_016878442.1

View Full Product Details