Product Details

SNP ID
rs973868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56796794 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCCCTCTCTCTGTCCCACCCCTG[C/T]GTGTTGCATTCCTCCCACCCCATGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf67 PubMed Links

Gene Details

Gene
C17orf67
Gene Name
chromosome 17 open reading frame 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085430.2 Intron NP_001078899.2
XM_011524735.2 Intron XP_011523037.1

View Full Product Details