Product Details

SNP ID
rs1504043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113890495 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCAGCCAGGGCCTCGCGGGTCCC[C/T]TCGTTTCTCCCTCCTGGGACTGGGG
Phenotype
MIM: 604222
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTR3 PubMed Links

Gene Details

Gene
ACTR3
Gene Name
ARP3 actin related protein 3 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277140.1 Intron NP_001264069.1
NM_005721.4 Intron NP_005712.1
Gene
LOC101060091
Gene Name
uncharacterized LOC101060091
There are no transcripts associated with this gene.

View Full Product Details