Product Details

SNP ID
rs1529805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:237798821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCGGAGAGCGGGGCGCTGGCC[C/T]GTGGAGGCGGCAGCGGCGGCGGCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM44 PubMed Links

Gene Details

Gene
RBM44
Gene Name
RNA binding motif protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080504.2 433 UTR 5 NP_001073973.2
XM_005246075.1 433 UTR 5 XP_005246132.1
XM_011511152.2 433 UTR 5 XP_011509454.1
XM_011511153.2 433 UTR 5 XP_011509455.1
XM_011511154.2 433 UTR 5 XP_011509456.1
XM_011511155.1 433 Intron XP_011509457.1
XM_017004054.1 433 UTR 5 XP_016859543.1
XM_017004055.1 433 UTR 5 XP_016859544.1
XM_017004056.1 433 Intron XP_016859545.1
XM_017004057.1 433 UTR 5 XP_016859546.1
XM_017004058.1 433 Intron XP_016859547.1

View Full Product Details