Product Details

SNP ID
rs1441093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233363838 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTGCTCTATGGGTTTGTAGTCCA[A/G]GAGCAATAGGTCATGCCATACAGAT
Phenotype
MIM: 601826
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKD PubMed Links

Gene Details

Gene
DGKD
Gene Name
diacylglycerol kinase delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003648.2 Intron NP_003639.2
NM_152879.2 Intron NP_690618.2
XM_005246107.3 Intron XP_005246164.1
XM_005246108.1 Intron XP_005246165.1
XM_005246110.3 Intron XP_005246167.1
XM_011512028.2 Intron XP_011510330.1
XM_011512029.2 Intron XP_011510331.1
XM_011512030.2 Intron XP_011510332.1
XM_011512031.2 Intron XP_011510333.1
XM_011512032.2 Intron XP_011510334.1
XM_011512033.2 Intron XP_011510335.1
XM_011512035.1 Intron XP_011510337.1
XM_011512037.2 Intron XP_011510339.1
XM_011512039.2 Intron XP_011510341.1
XM_011512040.2 Intron XP_011510342.1
XM_017005138.1 Intron XP_016860627.1
XM_017005139.1 Intron XP_016860628.1
XM_017005140.1 Intron XP_016860629.1

View Full Product Details