Product Details

SNP ID
rs1574424
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:196763491 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAAATAAAATGCATTTAAATTATA[C/T]ACATGGAGTATACAATATATTTACT
Phenotype
MIM: 604888
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTF3C3 PubMed Links

Gene Details

Gene
GTF3C3
Gene Name
general transcription factor IIIC subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206774.1 3890 Intron NP_001193703.1
NM_012086.4 3890 UTR 3 NP_036218.1
XM_005246965.4 3890 UTR 3 XP_005247022.1

View Full Product Details