Product Details

SNP ID
rs128473
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:21489594 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGCTCTGTCTGCTGCTTCCCTCTAA[G/T]CTGCCCATGGGGTGAGGACTCCTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PI4KAP2 PubMed Links

Gene Details

Gene
PI4KAP2
Gene Name
phosphatidylinositol 4-kinase alpha pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details