Product Details

SNP ID
rs1042347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:32388238 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATGGTTTTTATACACTAAGGACA[C/G]TCCCAGTGATGAAAATGAATTCCCC
Phenotype
MIM: 607102
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
WT1 PubMed Links

Gene Details

Gene
WT1
Gene Name
Wilms tumor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000378.4 1965 UTR 3 NP_000369.3
NM_001198551.1 1965 UTR 3 NP_001185480.1
NM_001198552.1 1965 UTR 3 NP_001185481.1
NM_024424.3 1965 UTR 3 NP_077742.2
NM_024426.4 1965 UTR 3 NP_077744.3

View Full Product Details