Product Details

SNP ID
rs831616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33701277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTCATCCACTACTGTGGGAGG[C/T]AGACAAAGGAGAAGAGGATTGATTG
Phenotype
MIM: 107271
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf91 PubMed Links

Gene Details

Gene
C11orf91
Gene Name
chromosome 11 open reading frame 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166692.1 Intron NP_001160164.1
XM_017017053.1 Intron XP_016872542.1
Gene
CD59
Gene Name
CD59 molecule
There are no transcripts associated with this gene.

View Full Product Details