Product Details

SNP ID
rs1057588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101006765 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATCTATCTATCTATCTGTAGCAT[C/T]TTAAAGGTAATGAAATTAATGTGGC
Phenotype
MIM: 603347
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPAS2 PubMed Links

Gene Details

Gene
NPAS2
Gene Name
neuronal PAS domain protein 2
There are no transcripts associated with this gene.

Gene
RPL31
Gene Name
ribosomal protein L31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000993.4 4899 Intron NP_000984.1
NM_001098577.2 4899 Intron NP_001092047.1
NM_001099693.1 4899 Intron NP_001093163.1
Gene
TBC1D8
Gene Name
TBC1 domain family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005263862.3 4899 UTR 3 XP_005263919.1
XM_005263863.4 4899 Intron XP_005263920.1
XM_005263864.3 4899 Intron XP_005263921.1
XM_011510507.1 4899 Intron XP_011508809.1
XM_011510508.2 4899 Intron XP_011508810.1
XM_017003198.1 4899 Intron XP_016858687.1
XM_017003199.1 4899 Intron XP_016858688.1
XM_017003200.1 4899 Intron XP_016858689.1
XM_017003201.1 4899 Intron XP_016858690.1
XM_017003202.1 4899 Intron XP_016858691.1

View Full Product Details