Product Details
- SNP ID
-
rs901474
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:112514888 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGCTGCTGCTCAGGGCGGCGCTGGC[G/T]CCTCCCGGGCTCAGCAACACTGCCT
- Phenotype
-
MIM: 606624
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NEUROG2
PubMed Links
Gene Details
- Gene
- NEUROG2
- Gene Name
- neurogenin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024019.3 |
915 |
Silent Mutation |
GGA,GGC |
G196G |
NP_076924.1 |
View Full Product Details