Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001303484.1 | Intron | NP_001290413.1 | |||
NM_031481.2 | Intron | NP_113669.1 | |||
XM_011546149.2 | Intron | XP_011544451.1 | |||
XM_011546150.2 | Intron | XP_011544452.1 | |||
XM_011546151.2 | Intron | XP_011544453.1 | |||
XM_011546152.1 | Intron | XP_011544454.1 | |||
XM_011546153.2 | Intron | XP_011544455.1 | |||
XM_011546154.2 | Intron | XP_011544456.1 | |||
XM_017028968.1 | Intron | XP_016884457.1 | |||
XM_017028969.1 | Intron | XP_016884458.1 | |||
XM_017028970.1 | Intron | XP_016884459.1 | |||
XM_017028971.1 | Intron | XP_016884460.1 | |||
XM_017028972.1 | Intron | XP_016884461.1 | |||
XM_017028973.1 | Intron | XP_016884462.1 | |||
XM_017028974.1 | Intron | XP_016884463.1 | |||
XM_017028975.1 | Intron | XP_016884464.1 |