Product Details
- SNP ID
-
rs1540179
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:77623894 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGTCTGGGGGAGGCAATCGGAAAG[A/G]GCTCTTAAATAAATGAAGTATGGTT
- Phenotype
-
MIM: 602158
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLNS1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115786252] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLNS1A
- Gene Name
- chloride nucleotide-sensitive channel 1A
- Gene
- LOC107987163
- Gene Name
- uncharacterized LOC107987163
There are no transcripts associated with this gene.
View Full Product Details