Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_016565.2 | Intron | NP_057649.2 | |||
XM_017017883.1 | Intron | XP_016873372.1 | |||
XM_017017884.1 | Intron | XP_016873373.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001267803.1 | Intron | NP_001254732.1 | |||
NM_001267804.1 | Intron | NP_001254733.1 | |||
NM_001267805.1 | Intron | NP_001254734.1 | |||
NM_001267806.1 | Intron | NP_001254735.1 | |||
NM_025155.2 | Intron | NP_079431.1 | |||
XM_005274308.2 | Intron | XP_005274365.2 | |||
XM_005274309.1 | Intron | XP_005274366.1 | |||
XM_017018357.1 | Intron | XP_016873846.1 | |||
XM_017018358.1 | Intron | XP_016873847.1 | |||
XM_017018359.1 | Intron | XP_016873848.1 |